CAS RN 6277-17-4
CAS RN 6277-17-4
IUPAC Name:
2-iodo-1-methyl-3-nitrobenzene
Synonyms:
2-iodo-1-methyl-3-nitrobenzene
Molecular Weight (g/mol):
263.03
Molecular Formula:
C7H6INO2
InChi Key:
AKYPSJARSQSCHJ-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(=C1I)[N+]([O-])=O
1
–
1
of
1
results
1
2-Iodo-3-nitrotoluene 98.0+%, TCI America™

CAS: 6277-17-4 Molecular Formula: C7H6INO2 Molecular Weight (g/mol): 263.034 MDL Number: MFCD00091040 InChI Key: AKYPSJARSQSCHJ-UHFFFAOYSA-N Synonym: 2-Iodo-1-methyl-3-nitrobenzene PubChem CID: 234781 IUPAC Name: 2-iodo-1-methyl-3-nitrobenzene SMILES: CC1=C(C(=CC=C1)[N+](=O)[O-])I