accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62778-16-9

IUPAC Name: diethyl [(2-methylphenyl)methyl]phosphonate
Synonyms: diethyl (2-methylphenyl)methylphosphonate
Molecular Weight (g/mol): 242.26
Molecular Formula: C12H19O3P
InChi Key: SAVIMLRIKAZZCZ-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=CC=C1C)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results