accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClICl

CAS RN 62778-19-2

IUPAC Name: 1,3-dichloro-2-iodo-5-nitrobenzene
Synonyms: 1,3-dichloro-2-iodo-5-nitrobenzene
Molecular Weight (g/mol): 317.89
Molecular Formula: C6H2Cl2INO2
InChi Key: FCDBEIIBODDDEJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(I)C(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results