accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 628-08-0

IUPAC Name: but-2-en-1-yl acetate
Synonyms: but-2-en-1-yl acetate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: WNHXJHGRIHUOTG-UHFFFAOYSA-N
SMILES: CC=CCOC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results