accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 628-13-7

IUPAC Name: pyridin-1-ium chloride
Synonyms: pyridium chloride
Molecular Weight (g/mol): 115.56
Molecular Formula: C5H6ClN
InChi Key: AOJFQRQNPXYVLM-UHFFFAOYSA-N
SMILES: [Cl-].C1=CC=[NH+]C=C1
Boiling Point 
  • (3)
  • (4)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (9)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Grade 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (3)
  • (6)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)
  • (9)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)