accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 628-67-1

IUPAC Name: 4-(acetyloxy)butyl acetate
Synonyms: 1,4-butanediol, diacetate
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: XUKSWKGOQKREON-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results