accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 628-94-4

IUPAC Name: hexanediamide
Synonyms: adipamide
Molecular Weight (g/mol): 144.17
Molecular Formula: C6H12N2O2
InChi Key: GVNWZKBFMFUVNX-UHFFFAOYSA-N
SMILES: NC(=O)CCCCC(N)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)