accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 628-99-9

IUPAC Name: nonan-2-ol
Synonyms: 2-nonanol
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: NGDNVOAEIVQRFH-UHFFFAOYNA-N
SMILES: CCCCCCCC(C)O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)