accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6280-80-4

IUPAC Name: 2-(2-formylphenoxy)acetic acid
Synonyms: 2-formylphenoxyacetic acid
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: ANWMNLAAFDCKMT-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results