accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOOCH3

CAS RN 6280-99-5

IUPAC Name: 1,4-dibutyl 2-hydroxybutanedioate
Synonyms: malic acid dibutyl ester
Molecular Weight (g/mol): 246.3
Molecular Formula: C12H22O5
InChi Key: PDSCSYLDRHAHOX-UHFFFAOYNA-N
SMILES: CCCCOC(=O)CC(O)C(=O)OCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results