CAS RN 62813-01-8
IUPAC Name:
1-cyclopropylpiperidin-4-one
Synonyms:
1-cyclopropylpiperidin-4-one
Molecular Weight (g/mol):
139.2
Molecular Formula:
C8H13NO
InChi Key:
DTUJRJIWGWTNFQ-UHFFFAOYSA-N
SMILES:
O=C1CCN(CC1)C1CC1
1
–
1
of
1
results
1
Promotions Available
1-Cyclopropyl-4-piperidone, 98%
1-Cyclopropyl-4-piperidone, 98%
CAS: 62813-01-8 Molecular Formula: C8H13NO Molecular Weight (g/mol): 139.20 MDL Number: MFCD07374375 InChI Key: DTUJRJIWGWTNFQ-UHFFFAOYSA-N Synonym: 1-cyclopropyl-4-piperidinone,1-cyclopropyl-4-piperidone,4-piperidinone, 1-cyclopropyl,1-cyclopropyl-4-oxopiperidine,4-oxopiperidin-1-yl cyclopropane,1-cyclopropyltetrahydro-4 1h-pyridinone,carbergoline,1-cyclopropyl-4-piperidon,n-cyclopropyl-4-piperidone,acmc-1b4f7 PubChem CID: 16227629 IUPAC Name: 1-cyclopropylpiperidin-4-one SMILES: O=C1CCN(CC1)C1CC1