CAS RN 6282-02-6
IUPAC Name:
N-(hydroxymethyl)benzamide
Synonyms:
benzamide, N-(hydroxymethyl)-
Molecular Weight (g/mol):
151.17
Molecular Formula:
C8H9NO2
InChi Key:
UOUBPDZUBVJZOQ-UHFFFAOYSA-N
SMILES:
OCNC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1