accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6282-88-8

HOCl

CAS RN 6282-88-8

IUPAC Name: 3-(4-chlorophenyl)propan-1-ol
Synonyms: 3-(4-chlorophenyl)propan-1-ol
Molecular Weight (g/mol): 170.64
Molecular Formula: C9H11ClO
InChi Key: ZHBIWFGGIKFSHZ-UHFFFAOYSA-N
SMILES: OCCCC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results