accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOOH

CAS RN 628297-55-2

IUPAC Name: 3-(1-methyl-1H-pyrazol-5-yl)benzoic acid
Synonyms: 3-(2-methylpyrazol-3-yl)benzoic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: OBOMYSVFLSMYLV-UHFFFAOYSA-N
SMILES: CN1N=CC=C1C1=CC(=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results