CAS RN 6284-46-4
CAS RN 6284-46-4
IUPAC Name:
propan-2-yl (2E)-but-2-enoate
Synonyms:
isopropyl (2E)-but-2-enoate
Molecular Weight (g/mol):
128.17
Molecular Formula:
C7H12O2
InChi Key:
AABBHSMFGKYLKE-SNAWJCMRSA-N
SMILES:
C\C=C\C(=O)OC(C)C
1
–
1
of
1
results
1
Isopropyl Crotonate 98.0+%, TCI America™
CAS: 6284-46-4 Molecular Formula: C7H12O2 Molecular Weight (g/mol): 128.171 MDL Number: MFCD00059053 InChI Key: AABBHSMFGKYLKE-SNAWJCMRSA-N Synonym: Crotonic Acid Isopropyl Ester PubChem CID: 5354359 IUPAC Name: propan-2-yl (E)-but-2-enoate SMILES: CC=CC(=O)OC(C)C