accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62871-09-4

IUPAC Name: 10-bromodec-1-ene
Synonyms: 10-bromodec-1-ene
Molecular Weight (g/mol): 219.17
Molecular Formula: C10H19Br
InChi Key: JVVPJOMYWVYPOF-UHFFFAOYSA-N
SMILES: BrCCCCCCCCC=C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)