accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNN

CAS RN 628711-58-0

IUPAC Name: 1-methyl-1H-indole-4-carbonitrile
Synonyms: 1-methylindole-4-carbonitrile
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: DYOVHBKIEBCIIE-UHFFFAOYSA-N
SMILES: CN1C=CC2=C(C=CC=C12)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results