accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH2OO

CAS RN 62873-16-9

IUPAC Name: 5-methyl-3-methylideneoxolan-2-one
Synonyms: 5-methyl-3-methylideneoxolan-2-one
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: KYLUHLJIAMFYKW-UHFFFAOYNA-N
SMILES: CC1CC(=C)C(=O)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results