accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62893-20-3

IUPAC Name: sodium (6R,7R)-7-[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetamido]-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: cefoperazone
Molecular Weight (g/mol): 667.65
Molecular Formula: C25H26N9NaO8S2
InChi Key: NCFTXMQPRQZFMZ-WERGMSTESA-M
SMILES: [Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SCC(CSC4=NN=NN4C)=C(N3C2=O)C([O-])=O)C2=CC=C(O)C=C2)C(=O)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Physical Form 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Type 
  • (4)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)