accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH

CAS RN 629-05-0

IUPAC Name: oct-1-yne
Synonyms: 1-octyne
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: UMIPWJGWASORKV-UHFFFAOYSA-N
SMILES: CCCCCCC#C
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (7)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (7)