accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 629-06-1

IUPAC Name: 1-chloroheptane
Synonyms: 1-chloroheptane
Molecular Weight (g/mol): 134.65
Molecular Formula: C7H15Cl
InChi Key: DZMDPHNGKBEVRE-UHFFFAOYSA-N
SMILES: CCCCCCCCl
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)