accessibility menu, dialog, popup

UserName

Viewing profile as user:
(Z)(Z)(Z)(Z)

CAS RN 629-20-9

IUPAC Name: (1Z,3Z,5Z,7Z)-cycloocta-1,3,5,7-tetraene
Synonyms: cyclooctatetraene
Molecular Weight (g/mol): 104.15
Molecular Formula: C8H8
InChi Key: KDUIUFJBNGTBMD-DLMDZQPMSA-N
SMILES: C1=C/C=C\C=C/C=C\1
Boiling Point 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)