accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 629-27-6

IUPAC Name: 1-iodooctane
Synonyms: 1-iodooctane
Molecular Weight (g/mol): 240.13
Molecular Formula: C8H17I
InChi Key: UWLHSHAHTBJTBA-UHFFFAOYSA-N
SMILES: CCCCCCCCI
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (4)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (6)
  • (4)
  • (3)
  • (3)
  • (4)
  • (4)
  • (3)
  • (3)
  • (5)