accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 629-40-3

IUPAC Name: octanedinitrile
Synonyms: octanedinitrile
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: BTNXBLUGMAMSSH-UHFFFAOYSA-N
SMILES: N#CCCCCCCC#N
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)