CAS RN 629-56-1
IUPAC Name:
hexadec-2-enoic acid
Synonyms:
t-2-hexadecenoic acid
Molecular Weight (g/mol):
254.41
Molecular Formula:
C16H30O2
InChi Key:
ZVRMGCSSSYZGSM-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCC=CC(O)=O
1
–
1
of
1
results
1
2-Hexadecenoic Acid 98.0+%, TCI America™
CAS: 629-56-1 Molecular Formula: C16H30O2 Molecular Weight (g/mol): 254.414 MDL Number: MFCD00045907 InChI Key: ZVRMGCSSSYZGSM-CCEZHUSRSA-N Synonym: Gaidic Acid PubChem CID: 5282743 ChEBI: CHEBI:37252 IUPAC Name: (E)-hexadec-2-enoic acid SMILES: CCCCCCCCCCCCCC=CC(=O)O