accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 629-58-3

IUPAC Name: pentadecyl acetate
Synonyms: pentadecyl acetate
Molecular Weight (g/mol): 270.46
Molecular Formula: C17H34O2
InChi Key: AAAIZLQILBFRTJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results