accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 629-65-2

IUPAC Name: 1-(heptylsulfanyl)heptane
Synonyms: heptane, 1,1'-thiobis-
Molecular Weight (g/mol): 230.45
Molecular Formula: C14H30S
InChi Key: LEMIDOZYVQXGLI-UHFFFAOYSA-N
SMILES: CCCCCCCSCCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results