accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 629-83-4

IUPAC Name: methyl triacontanoate
Synonyms: methyl triacontanoate
Molecular Weight (g/mol): 466.84
Molecular Formula: C31H62O2
InChi Key: BIRUBGLRQLAEFF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results