CAS RN 629-90-3
CAS RN 629-90-3
IUPAC Name:
heptadecanal
Synonyms:
heptadecanal
Molecular Weight (g/mol):
254.46
Molecular Formula:
C17H34O
InChi Key:
PIYDVAYKYBWPPY-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCC=O
1
–
1
of
1
results
1
Heptadecanal 97.0+%, TCI America™
CAS: 629-90-3 Molecular Formula: C17H34O Molecular Weight (g/mol): 254.458 MDL Number: MFCD04034658 InChI Key: PIYDVAYKYBWPPY-UHFFFAOYSA-N Synonym: Heptadecanaldehyde, Heptadecyl Aldehyde PubChem CID: 71552 IUPAC Name: heptadecanal SMILES: CCCCCCCCCCCCCCCCC=O