accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CI

CAS RN 629-93-6

IUPAC Name: 1-iodooctadecane
Synonyms: 1-iodooctadecane
Molecular Weight (g/mol): 380.4
Molecular Formula: C18H37I
InChi Key: ZNJOCVLVYVOUGB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCI
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)