accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 629-99-2

IUPAC Name: pentacosane
Synonyms: pentacosane
Molecular Weight (g/mol): 352.69
Molecular Formula: C25H52
InChi Key: YKNWIILGEFFOPE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)