accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62942-43-2

IUPAC Name: (2-oxoethyl)triphenylphosphanium chloride
Synonyms: (2-oxoethyl)triphenylphosphanium chloride
Molecular Weight (g/mol): 340.79
Molecular Formula: C20H18ClOP
InChi Key: RVEJRPJGKXTQIF-UHFFFAOYSA-M
SMILES: [Cl-].O=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)