accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOSSN

CAS RN 6295-57-4

IUPAC Name: 2-(1,3-benzothiazol-2-ylsulfanyl)acetic acid
Synonyms: (1,3-benzothiazol-2-ylsulfanyl)acetic acid
Molecular Weight (g/mol): 225.28
Molecular Formula: C9H7NO2S2
InChi Key: ZZUQWNYNSKJLPI-UHFFFAOYSA-N
SMILES: OC(=O)CSC1=NC2=CC=CC=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results