accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6295-87-0

IUPAC Name: 1-aminopyridin-1-ium iodide
Synonyms: 1-aminopyridin-1-ium iodide
Molecular Weight (g/mol): 222.03
Molecular Formula: C5H7IN2
InChi Key: NDRLPYIMWROJBG-UHFFFAOYSA-M
SMILES: [I-].N[N+]1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)