accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6296-53-3

IUPAC Name: N-(1,3-dioxo-1,3-dihydro-2-benzofuran-4-yl)acetamide
Synonyms: N-(1,3-dioxo-2-benzofuran-4-yl)acetamide
Molecular Weight (g/mol): 205.17
Molecular Formula: C10H7NO4
InChi Key: PAUAJOABXCGLCN-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC2=C1C(=O)OC2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results