accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OHOHNCH3CH3OCH3CH3CH3O

CAS RN 62965-35-9

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3,3-dimethylbutanoic acid
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: LRFZIPCTFBPFLX-JLDDOWRYNA-N
SMILES: CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C
Molecular Weight (g/mol) 
  • (8)
  • (1)
  • (1)
Quantity 
  • (2)
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (6)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (6)
  • (8)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (2)
  • (6)
  • (1)