accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6298-11-9

IUPAC Name: 2-(pyridin-4-yl)ethane-1-thiol hydrochloridyl
Synonyms: 2-(pyridin-4-yl)ethanethiol hydrochloridyl
Molecular Weight (g/mol): 174.67
Molecular Formula: C7H9ClNS
InChi Key: HXMNVHPGTRZNSP-UHFFFAOYSA-N
SMILES: [Cl].SCCC1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)