accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6299-67-8

IUPAC Name: 2,3-dimethoxyaniline
Synonyms: 2,3-dimethoxyaniline
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: HEZIOZBMPKPOER-UHFFFAOYSA-N
SMILES: COC1=CC=CC(N)=C1OC
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results