CAS RN 63-05-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Androstenedione Solution, 1.0 mg/mL in acetonitrile, Sold by MilliporeSigma™ Supelco™
Suitable for use as starting material in linearity standards, calibrators, and controls for numerous LC/MS and GC/MS applications in endocrinology, clinical chemistry, neonatal screening, and sports testing.