accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 630-22-8

IUPAC Name: 2,2-dimethylpropanethioamide
Synonyms: 2,2-dimethylpropanethioamide
Molecular Weight (g/mol): 117.21
Molecular Formula: C5H11NS
InChi Key: FJZJUSOFGBXHCV-UHFFFAOYSA-N
SMILES: CC(C)(C)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results