accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 630-88-6

IUPAC Name: 3',6'-dichloro-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 3',6'-dichlorospiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 369.2
Molecular Formula: C20H10Cl2O3
InChi Key: LNBKJWCRFAZVLK-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(OC3=CC(Cl)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results