accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63010-72-0

IUPAC Name: 4-chloro-8-fluoroquinoline
Synonyms: 4-chloro-8-fluoroquinoline
Molecular Weight (g/mol): 181.59
Molecular Formula: C9H5ClFN
InChi Key: NUIMKHDPMIPYFI-UHFFFAOYSA-N
SMILES: FC1=C2N=CC=C(Cl)C2=CC=C1
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)