accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6304-39-8

IUPAC Name: octanehydrazide
Synonyms: octanoic acid hydrazide
Molecular Weight (g/mol): 158.25
Molecular Formula: C8H18N2O
InChi Key: VMUZVGRNTPFTKE-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)NN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results