accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOH2NNH2

CAS RN 63069-49-8

IUPAC Name: 2-amino-5-fluorobenzamide
Synonyms: 2-amino-5-fluorobenzamide
Molecular Weight (g/mol): 154.14
Molecular Formula: C7H7FN2O
InChi Key: RHJMYIPLYKQZJM-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N)C=CC(F)=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results