accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6307-83-1

IUPAC Name: 3-bromo-5-nitrobenzoate
Synonyms: 3-bromo-5-nitrobenzoate
Molecular Weight (g/mol): 245.01
Molecular Formula: C7H3BrNO4
InChi Key: AXRKIZCFYZBBPX-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(Br)=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results