accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FO

CAS RN 63082-45-1

IUPAC Name: 4-fluoro-2-methylbenzaldehyde
Synonyms: 4-fluoro-2-methylbenzaldehyde
Molecular Weight (g/mol): 138.14
Molecular Formula: C8H7FO
InChi Key: ADCFIKGEGWFWEA-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results