accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)NH3S

CAS RN 6309-16-6

IUPAC Name: (1R)-1-(thiophen-2-yl)ethan-1-aminium
Synonyms: (1R)-1-(thiophen-2-yl)ethanaminium
Molecular Weight (g/mol): 128.21
Molecular Formula: C6H10NS
InChi Key: LYJBVRVJQXVVPI-RXMQYKEDSA-O
SMILES: C[C@@H]([NH3+])C1=CC=CS1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)