accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63094-81-5

IUPAC Name: tert-butyl 2-amino-3-carbamoylpropanoate hydrochloride
Synonyms: tert-butyl 2-amino-3-carbamoylpropanoate hydrochloride
Molecular Weight (g/mol): 224.69
Molecular Formula: C8H17ClN2O3
InChi Key: RXNKCUXXNGWROA-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)OC(=O)C(N)CC(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)