accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2S

CAS RN 631-58-3

IUPAC Name: propanethioamide
Synonyms: propanethioamide
Molecular Weight (g/mol): 89.16
Molecular Formula: C3H7NS
InChi Key: WPZSAUFQHYFIPG-UHFFFAOYSA-N
SMILES: CCC(N)=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results