CAS RN 631-61-8
Ammonium Acetate (Optima LC/MS), Fisher Chemical
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Ammonium acetate, 97%
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 MDL Number: MFCD00013066 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Ammonium acetate, 5M aq. soln.
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 MDL Number: MFCD00013066 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Acetic acid, ammonium salt, 98+%, for HPLC
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Ammonium acetate, ACS, 97.0% min
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 MDL Number: MFCD00013066 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Ammonium Acetate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 MDL Number: MFCD00013066 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Ammonium Acetate (Crystalline/HPLC), Fisher Chemical
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 MDL Number: MFCD00013066 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Acetic acid, ammonium salt, 98%, pure
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.08 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
MP Biomedicals™ Ammonium Acetate, Ultra Pure Grade
Ammonium Acetate, Ultra Pure Grade is used in the purification and precipitation of DNA and for protein crystallization.
Ammonium Acetate, ∽99%, (Crystalline), MP Biomedicals
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Acetic acid, ammonium salt, 97+%, ACS reagent
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.08 MDL Number: MFCD00013066 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Ammonium Acetate, ∽98%, MP Biomedicals
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Thermo Scientific Chemicals Acetic acid, ammonium salt, 98%, for biochemistry
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Ammonium Acetate, Extra Pure, MilliporeSigma™
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]
Ammonium acetate, ∽98%, MP Biomedicals™
CAS: 631-61-8 Molecular Formula: C2H7NO2 Molecular Weight (g/mol): 77.083 InChI Key: USFZMSVCRYTOJT-UHFFFAOYSA-N Synonym: ammonium acetate,acetic acid, ammonium salt,azanium acetate,ammoniumacetate,acetic acid ammonium salt,ammonium ethanoate,unii-rre756s6q2,aconh4,ch3coonh4,ch3co2nh4 PubChem CID: 517165 ChEBI: CHEBI:62947 IUPAC Name: azanium;acetate SMILES: CC(=O)[O-].[NH4+]